Idex® Frit-in-a-Ferrule™ Filtration System
Combine sealing and filtering in one fitting assembly
  • Save money and time by eliminating separate filter fittings
  • Use a flat bottom 1/4-28, M6, or 5/16-24 connection
The frit-in-a-ferrule design uses a flat-bottom ferrule with a frit inside the body to filter and seal simultaneously. This reduces the number of connections in a system by eliminating the need for separate in-line filter assemblies.
3,300.00 - ₹57,200.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Diameter (mm) Pore Size (µm) Membrane Material
Availability Pricing
Idex Frit-In-A-Ferrule, Super Flangeless, Natural, SS Frit, for 1/16" OD Tubing 4271168 Mfr # P-270
COMPARE
0.062 2 Stainless Steel
₹4,000.00INR / Each
Idex Frit-In-A-Ferrule, Flangeless, Green, SS/PCTFE, 2 µm; 1/EA 4271169 Mfr # P-272
COMPARE
0.062 2 Stainless Steel
₹3,400.00INR / Each
Idex Frit-In-A-Ferrule, Flangeless, Black, SS/PCTFE, 0.5 µm; 1/EA 4271170 Mfr # P-273
COMPARE
0.062 0.5 Stainless Steel
₹4,000.00INR / Each
Idex Frit-In-A-Ferrule, Super Flangeless, Natural, PEEK Frit, for 1/16" OD Tubing 4271171 Mfr # P-274
COMPARE
0.046 2 PEEK
₹4,500.00INR / Each
Idex Frit-In-A-Ferrule, Super Flangeless, Natural, ETFE, SS lock ring, 10 µm; 1/EA 4271173 Mfr # P-276
COMPARE
0.062 10 Stainless Steel
₹4,100.00INR / Each
Idex Frit-In-A-Ferrule, Super Flangeless, Natural, SS Frit, for 1/8" OD Tubing 4271174 Mfr # P-370
COMPARE
0.062 2 Stainless Steel
₹4,100.00INR / Each
Idex Frit-In-A-Ferrule, Super Flangeless, Black, PEEK, SS lock ring, 0.5 µm; 10/pk 4271175 Mfr # P-371X
COMPARE
0.062 0.5 Stainless Steel
₹57,200.00INR / Pkg of 10
Idex Frit-In-A-Ferrule, Flangeless, Green, PCTFE & SS, 2 µm; 1/EA 4271176 Mfr # P-372
COMPARE
0.062 2 Stainless Steel
₹3,800.00INR / Each
Idex Frit-In-A-Ferrule, Super Flangeless, Black, SS/PCTFE, 0.5 µm; 1/EA 4271177 Mfr # P-373
COMPARE
0.062 0.5 Stainless Steel
₹3,300.00INR / Each
Idex Frit-In-A-Ferrule, Super Flangeless, Natural, PEEK Frit, for 1/8" OD Tubing 4271178 Mfr # P-374
COMPARE
0.062 2 PEEK
₹4,400.00INR / Each
Showing 1- 10 of 10