Idex® Flangeless Short Head Fitting Assemblies, 1/4-28 Flat-Bottom for 1/8" OD Tubing
Short head is ideal for tight-speace applications
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
2,500.00 - ₹16,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Idex Flangeless Fitting, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0201504 Mfr # XP-335X
COMPARE
Natural 1/8 PEEK
₹16,800.00INR / Pkg of 10
Idex Flangeless Fitting, Short Head, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202096 Mfr # XP-318
COMPARE
Natural 1/8 ETFE
₹4,900.00INR / Each
Idex Flangeless Fitting, Short Head, Natural PEEK, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/Ea 0202099 Mfr # XP-335
COMPARE
Natural 1/8 PEEK
₹2,500.00INR / Each