Idex® Flangeless Ferrules, 5/16-24 Flat-Bottom Connection for 4 mm OD Tubing
Excellent for connecting rigid and semi-rigid tubing
  • Excellent for connecting rigid or semi-solid tubing
Use with male nuts for a leak-tight connection.
2,400.00 - ₹23,400.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (mm) Fitting Material Thread size UNF(M) (port 2) (InchShort)
Availability Pricing
Idex Flangeless Ferrule, Natural ETFE, 4 mm OD Tubing, 5/16-24 Flat-Bottom; 1/EA 0202035 Mfr # P-344
COMPARE
4 ETFE 5/16-24 flat bottom
₹2,400.00INR / Each
Idex Flangeless Ferrule, Natural ETFE, 4 mm OD Tubing, 5/16-24 Flat-Bottom; 10/PK 0202036 Mfr # P-344X
COMPARE
4 ETFE 5/16-24 flat bottom
₹23,400.00INR / Pkg of 10