Idex® Flangeless Ferrules, 1/4-28 or M6 Flat-Bottom Connection, ETFE

No need to flange tubing
  • Excellent for connecting rigid and semi-rigid tubing
Use with male nuts for a leak-tight connection.
490.00 - ₹4,800.00INR / Pkg of 10

7 variations of this product are available.

See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (mm) Description
Availability Pricing
Idex Flangeless Ferrule, Green ETFE, 1.8 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 10/PK 0201383 Mfr # P-342X
COMPARE
Green 1.8 Flangeless Ferrule, Green ETFE, 1.8 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 10/PK
₹4,700.00INR / Pkg of 10
Idex Flangeless Ferrule, Green ETFE, 1.8 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 1/EA 0202032 Mfr # P-342
COMPARE
Green 1.8 Flangeless Ferrule, Green ETFE, 1.8 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 1/EA
₹490.00INR / Each
Idex Flangeless Ferrule, Orange ETFE, 3.0 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 1/EA 0202033 Mfr # P-343
COMPARE
Orange 3 Flangeless Ferrule, Orange ETFE, 3.0 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 1/EA
₹490.00INR / Each
Idex Flangeless Ferrule, Orange ETFE, 3.0 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 10/PK 0202034 Mfr # P-343X
COMPARE
Orange 3 Flangeless Ferrule, Orange ETFE, 3.0 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 10/PK
₹4,800.00INR / Pkg of 10
Idex Flangeless Ferrule, White ETFE, 2.0 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 10/PK 0202044 Mfr # P-353X
COMPARE
White 2 Flangeless Ferrule, White ETFE, 2.0 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 10/PK
₹4,600.00INR / Pkg of 10
Idex Flangeless Ferrule, Red ETFE, 2.0 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 1/EA 0202045 Mfr # P-363R
COMPARE
Red 2 Flangeless Ferrule, Red ETFE, 2.0 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 1/EA
₹490.00INR / Each
Idex Flangeless Ferrule, Red ETFE, 2.0 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 10/PK 0202046 Mfr # P-363RX
COMPARE
Red 2 Flangeless Ferrule, Red ETFE, 2.0 mm OD Tubing, 1/4-28 or M6 Flat-Bottom; 10/PK
₹4,600.00INR / Pkg of 10
Showing 1- 7 of 7

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat