IC Biomedical HC Series High-Capacity Liquid Nitrogen Dewars
Engineered for maximum capacity and efficiency, the HC Series Freezers offer secure, long-term cryogenic storage without compromise
  • High-strength ribbed aluminum body with magneformed necktube for exceptional durability
  • Numbered index ring and tapered canister locator for organized, easy access
  • Superior vacuum insulation delivers extended holding times and thermal performance
  • Lockable lid provides enhanced security for sensitive samples
The HC Series High-Capacity Dewars are designed to meet the demands of long-term cryogenic storage where space, security, and performance matter most. Featuring a rugged ribbed aluminum body and a magneformed necktube for added strength, these freezers ensure reliable structural integrity. An intuitive numbered index ring and tapered canister locator streamline organization and retrieval. With advanced super insulation for superior vacuum performance and a lockable lid for sample protection, the HC Series delivers peace of mind and maximum efficiency in any cryogenic storage application.
272,000.00 - ₹343,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (Liters) Static Holding Time (days)
Availability Pricing
IC Biomedical 20HC-11M Cryogenic Storage Dewar System, 20 L, 6 Canisters 0377912 Mfr # 20HC-11M
COMPARE
20 87
₹272,000.00INR / Each
IC Biomedical 35HCB-11M Cryogenic Storage Dewar System, 35 L, 10 Canisters 0377941 Mfr # 35HCB-11M
COMPARE
35 130
₹343,000.00INR / Each