Huber KISS Refrigerated Heating Baths
Economic solution for sample temperature control
  • Natural refrigerants for environmentally friendly temperature control
  • Pump connections for externally attached applications
  • Overtemperature and low level protection
  • Safety Class III/FL (DIN 12876)
These Refrigerated Heating Baths are ideal for a variety of heating and cooling tasks in research and industry such as sample temperature control, materials testing, quality control, analyses, etc. These rugged units consist of a stainless steel bath, immersion thermostat, pressure and suction pump with wetted materials made from industrial plastic or stainless steel, and air cooled refrigeration unit. In addition, the cooling circulators include automatic refrigeration capacity adjustment which reduces energy consumption and waste heat down to the absolute essential. New KISS controller combines state-of-the-art technology with stylish design. The simple to navigate menu makes operating these baths as easy as pushing a few buttons. A large OLED display simultaneously shows set point, internal temperature, minimum temperature, maximum temperature, and status display of pump, cooling, and heating. Other features include USB and RS232 interfaces, autostart function if power is lost, overtemperature and low level protection.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity At 0° C (watts) Max Flow Rate (L/min) Power (VAC)
Availability Pricing
Huber KISS K15  Refrigerated Heating Bath, 115VAC, 60Hz WW-12751-34 Mfr # 2010.0018.98
COMPARE
200 14 115
Call For Price
Huber KISS K6  Refrigerated Heating Bath, 110 VAC, 50/60Hz WW-12751-37 Mfr # 2008.0045.98
COMPARE
150 14 110