Hoffman Stainless Steel Junction Boxes with Continuous-Hinge and Clamps, Type 4X
Protects components from indoor or outdoor corrosive environments
  • Seams continuously welded and ground smooth
  • Seamless foam-in-place gasket
  • Features stainless steel screws and clamps
  • Bonding provision on door and body
  • Ratings: NEMA Type 4X, IP66
Used in either indoor or outdoor applications, these enclosures combine a rugged continuous hinge, seamless foam-in-place gasket and stainless steel screw-down clamps for a reliable seal that protects components from corrosive environments. The cover and sides of the body have a smooth #4 brushed finish.
208,000.00 - ₹529,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
nVent Hoffman A1212CHNFSS Stainless Steel Junction Box, Continuous Hinge, type 4X 1050223 Mfr # A1212CHNFSS
COMPARE
6 12 12
₹318,000.00INR / Each
nVent Hoffman A1614CHNFSS6 Stainless Steel Junction Box, Continuous Hinge, type 4X 1050224 Mfr # A1614CHNFSS6
COMPARE
6 16 14
₹529,000.00INR / Each
nVent Hoffman A1008CHNFSS Stainless Steel Junction Box, Continuous Hinge, type 4X 1050268 Mfr # A1008CHNFSS
COMPARE
4 10 8
₹208,000.00INR / Each
nVent Hoffman A12106CHNFSS Stainless Steel Junction Box, Continuous Hinge, type 4X 1050269 Mfr # A12106CHNFSS
COMPARE
6 12 10
₹290,000.00INR / Each