Hoffman QLINE I Polycarbonate Wall-Mount Enclosures, Type 4X
Protects larger controls from corrosive environments
  • Body is of halogen-free, impact-resistant polycarbonate
  • Solid and clear window covers are of halogen-free, impact-resistant polycarbonate
  • Polycarbonate material is easily punched, drilled, filed or sawed
  • Mounting holes are molded directly under cover screws
  • Features molded internal pads for mounting optional panels, rails and other components (call for details); screws included
  • Features molded internal rails for mounting optional adjustable-depth panel kit (call for details)
  • Cover includes a molded-in hole for wire lead
  • Seamless foam-in-place gasket ensures a water- and dust-tight seal
  • Ratings: NEMA Type 4X, IP66
The QLINE I series enclosures provide easy access and protect larger controls from corrosive environments. These screw-cover enclosures have easy-to-remove covers attached to body with strong, durable polyamide cross-point cover screws. The polycarbonate enclosure material features a RAL 7035 light gray finish inside and out.
54,300.00 - ₹71,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
nVent Hoffman Q302013PCICC QLINE Wall-Mount Enclosure I type 4X Polycarbonate 1050336 Mfr # Q302013PCICC
COMPARE
₹54,300.00INR / Each
nVent Hoffman Q403013PCICC QLINE Wall-Mount Enclosure I type 4X Polycarbonate 1050339 Mfr # Q403013PCICC
COMPARE
₹71,900.00INR / Each