Hoffman QLINE D Polycarbonate Pushbutton Wall-Mount Enclosures, Type 4X and ABS Wall-Mount Enclosures
House and insulate 30.5-mm or 22.5-mm pushbuttons in hostile environments
  • High-impact polycarbonate is used for both body and cover; material is easily punched, drilled, filed or sawed
  • Straight side walls permit ganging of multiple enclosures
  • Seamless foam-in-place gasket ensures a water- and dust-tight seal
  • Material is halogen free
  • Cross-point captivated cover screws are stainless steel; protected by removable tamper-resistant caps
  • Mounting holes molded directly adjacent to cover screws
  • Molded internal pads for mounting optional panels, rails or other components (call for details)
  • Ratings: NEMA Type 4X, IP67, IEC 60529
QLINE™ D Enclosures have contoured bodies with flush cover screws for an attractive, contemporary appearance. Polycarbonate enclosure material is RAL 7035 light gray inside and out. Optional panels are unpainted plated steel.
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
nVent Hoffman Q1PBPCDM QLINE D Wall-Mount Enclosure PC NEMA 4X, 75x73x79mm, 22.5mm Hole 1050331 Mfr # Q1PBPCDM
COMPARE
3.23 3.15
₹23,400.00INR / Each
nVent Hoffman Q2PBPCDM QLINE D Wall-Mount Enclosure PC NEMA 4X, 113x73x79mm, 22.5mm Hole 1050334 Mfr # Q2PBPCDM
COMPARE
4.72 3.15
₹24,800.00INR / Each