Hoffman INLINE 304 Stainless Steel Wall-Mount Instrumentation Enclosures, Type 4X
Outward formed body flange prevents contaminants from entering the enclosure
  • 16 gauge stainless steel covers and doors and 14 gauge bodies
  • Seams continuously welded and ground smooth
  • External formed body flange trough
  • Large perimeter body opening
  • Seamless foam-in-place gasket
  • Hinged-door enclosure has removable 316 stainless steel hinge pins and self-grounding, slotted, quarter-turn 316 stainless steel latch system with double seal
  • Screw-cover enclosure has captivated 316 stainless steel cover screws with sealing bushings; internal compression stops maintain optimal gasket compression
  • Fastening holes in back of body for direct mounting or optional external mounting brackets (call for details)
  • M6 bonding provision on cover; M6 ground stud in body
  • Ratings: NEMA Type 4X, IP66, IEC 60529
Aesthetic styling, flush latches and an attractive, stroked finish make INLINE™ Enclosures an excellent choice when style and functionality are important.
Cover and body have smooth #4 brushed finish.
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
nVent Hoffman LSC302515SS INLINE 304 SS NEMA 4X Enclosure, Hinged-Cover, 300x250x150mm 1050233 Mfr # LSC302515SS
COMPARE
5.91 9.84 11.81
₹287,000.00INR / Each