Hoffman Continuous-Hinge Fiberglass Wall-Mount Enclosures, Type 4X
Durable design and construction for corrosive environments
  • Compression-molded fiberglass offers outstanding chemical and temperature resistance and excellent weatherability
  • Light gray fiberglass is easily punched, drilled, filed or sawed
  • Continuous hinge and gasket ensure a secure water- and dust-tight seal
  • Polyester mounting brackets and stainless steel attachment screws are provided with each enclosure
  • Scratch-resistant polycarbonate windows are permanently bonded in place
  • Molded-in-place threaded brass inserts and plated steel screws are provided for mounting optional panels and terminal block kits
  • Removable hinged cover attached to body with Type 316 stainless steel hinge pin
  • Hinge pin and bail are corrosion-resistant type 316 stainless steel
  • Ratings: NEMA Type 4X, IP66
Fiberglass material is light-gray inside and out. Optional steel panels are painted white. Optional stainless steel, aluminum, conductive and composite panels are unpainted.
39,500.00 - ₹147,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
nVent Hoffman A16148CHSCFG Hinged Fiberglass NEMA 4X Enclosure, Solid/Quick, 16x14x8" 1050045 Mfr # A16148CHSCFG
COMPARE
₹90,900.00INR / Each
nVent Hoffman A664CHSCFG Hinged Fiberglass NEMA 4X Enclosure, Solid/Screw, 6x6x4" 1050057 Mfr # A664CHSCFG
COMPARE
₹39,500.00INR / Each
nVent Hoffman A664CHSCFGW Hinged Fiberglass NEMA 4X Enclosure, Window/Screw, 6x6x4" 1050058 Mfr # A664CHSCFGW
COMPARE
₹57,200.00INR / Each
nVent Hoffman A864CHSCFG Hinged Fiberglass NEMA 4X Enclosure, Solid/Screw, 8x6x4" 1050061 Mfr # A864CHSCFG
COMPARE
₹41,500.00INR / Each
nVent Hoffman A1086CHQRFG Hinged Fiberglass NEMA 4X Enclosure, Solid/Quick, 10x8x6" 1050243 Mfr # A1086CHQRFG
COMPARE
₹58,500.00INR / Each
nVent Hoffman A1086CHQRFGW Hinged Fiberglass NEMA 4X Enclosure, Window/Quick, 10x8x6" 1050244 Mfr # A1086CHQRFGW
COMPARE
₹93,100.00INR / Each
nVent Hoffman A12106CHQRFG Hinged Fiberglass NEMA 4X Enclosure, Solid/Quick, 12x10x6" 1050245 Mfr # A12106CHQRFG
COMPARE
Call For Price
nVent Hoffman A12106CHSCFG Hinged Fiberglass NEMA 4X Enclosure, Solid/Screw, 12x10x6" 1050247 Mfr # A12106CHSCFG
COMPARE
₹59,400.00INR / Each
nVent Hoffman A16148CHQRFG Hinged Fiberglass NEMA 4X Enclosure, Solid/Quick, 16x14x8" 1050250 Mfr # A16148CHQRFG
COMPARE
₹100,000.00INR / Each
nVent Hoffman A16148CHQRFGW Hinged Fiberglass NEMA 4X Enclosure, Window/Quick, 16x14x8" 1050251 Mfr # A16148CHQRFGW
COMPARE
₹147,000.00INR / Each
nVent Hoffman A181610CHQRFG Hinged Fiberglass NEMA 4X Enclosure, Solid/Quick, 18x16x10" 1050252 Mfr # A181610CHQRFG
COMPARE
₹139,000.00INR / Each
nVent Hoffman A664CHQRFG Hinged Fiberglass NEMA 4X Enclosure, Solid/Quick, 6x6x4" 1050253 Mfr # A664CHQRFG
COMPARE
₹50,700.00INR / Each
Showing 1- 12 of 12