Hirschmann® opus® Titrators
Obtain precise titration results with electronic control and dispensing
  • Configure titration parameters on user-friendly TFT touch screen
  • Time-saving rapid titration in conjunction with fine titration ensures system accuracy
  • Save up to nine procedures and settings with on-board memory for quick recall
  • Easily transfer data via RS-232 and USB interfaces
The electronic titration system is user-friendly with an intuitive touch screen and direct titration from the laboratory bottle. Titration impulse triggered by the motor allows for droplet titration of 1 droplet per button push. Prevent reagent loss with simple and rapid venting, made possible with the media recirculating system.
Advanced safety and protection features are valuable for daily routines. Protect your data and procedures from unauthorized personnel using the PIN function. The PIN function can also enable only select functions to be performed by selected users. Freely rotate the titrator on the reagent bottle, to ensure the bottle label is always pointing towards the user. Your light-sensitive samples will be protected by the light protection window, included with your titration system.
751,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Dispensing Accuracy Description
Availability Pricing
Hirschmann opus Titrator, 20 mL; 100 to 240 VAC, 50/60 Hz 2550305 Mfr # 9582020
COMPARE
20 R=0.2%; CV=0.07% Titrator, 20 mL; 100 to 240 VAC, 50/60 Hz
₹751,000.00INR / Each
Hirschmann opus Titrator, 50 mL; 100 to 240 VAC, 50/60 Hz 2550306 Mfr # 9582050
COMPARE
50 R=0.2%; CV=0.05% Titrator, 50 mL; 100 to 240 VAC, 50/60 Hz
₹751,000.00INR / Each