Hi-Lo Temperature Tape
Fiberglass tape is ideal for labeling cryogenic samples, ovens, and sterilizers; or for any application that involves extreme temperatures. Withstands temperatures from –385 to 550°F
(–232 to 288°C). Textured surface can be stamped or written on with a pencil.
5,600.00 - ₹11,000.00INR / Roll of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Width (in)
Availability Pricing
COMPARE
25 1/2
₹5,600.00INR / Roll of 1
COMPARE
25 1
₹11,000.00INR / Roll of 1