HEMCO Fume Hood Work Surfaces
Durable fume hood work surfaces handle the toughest applications
  • Designed to suit the specific needs of your lab, ranging from surfaces built for aggressive applications to general use
  • Constructed to fit Hemco fume hoods
  • Meets UL 1805 classification and high heat requirement
The fume hood work surfaces are available in either phenolic or epoxy resin.
265,000.00 - ₹1,243,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Depth (in) Exterior Width (in) Description
Availability Pricing
HEMCO Phenolic Work Surface for Universal and Clean Aire II, flat, 30" W 3370684 Mfr # 93028
COMPARE
23 30 Phenolic Work Surface for Universal and Clean Aire II, flat, 30" W
₹265,000.00INR / Each
HEMCO Phenolic Work Surface for UniFlow CE AireStream Fume Hoods, flat, 30" W 3370691 Mfr # 13018
COMPARE
24 30 Phenolic Work Surface for UniFlow CE AireStream Fume Hoods, flat, 30" W
Call For Price
HEMCO Phenolic Work Surface for UniFlow CE AireStream Fume Hoods, flat, 36" W 3370692 Mfr # 13618
COMPARE
24 36 Phenolic Work Surface for UniFlow CE AireStream Fume Hoods, flat, 36" W
Call For Price
HEMCO Phenolic Work Surface for UniFlow CE AireStream Fume Hoods, flat, 48" W 3370693 Mfr # 14818
COMPARE
24 48 Phenolic Work Surface for UniFlow CE AireStream Fume Hoods, flat, 48" W
₹416,000.00INR / Each
HEMCO Epoxy Resin Work Surface for UniFlow LE AireStream Fume Hoods, 48" W 3370696 Mfr # 20415
COMPARE
30 48 Epoxy Resin Work Surface for UniFlow LE AireStream Fume Hoods, 48" W
₹1,243,000.00INR / Each
HEMCO Epoxy Resin Work Surface for UniFlow LE AireStream Fume Hoods, 72" W 3370698 Mfr # 20615
COMPARE
30 72 Epoxy Resin Work Surface for UniFlow LE AireStream Fume Hoods, 72" W
Call For Price
HEMCO Epoxy Resin Work Surface for UniFlow LE AireStream Fume Hoods, 96" W 3370699 Mfr # 20815
COMPARE
30 96 Epoxy Resin Work Surface for UniFlow LE AireStream Fume Hoods, 96" W
Call For Price
Showing 1- 7 of 7