Heavy-Duty Tubing Clamps
Rugged and versatile tubing clamps for precise flow control
  • Designed to withstand elevated working pressures
  • Fast and secure installation with a fully adjustable swing-away locking screw
  • Compatible with silicone, braided silicone, and TPE tubing up to 1 inch OD
  • Lightweight and designed for easy single-hand operation
Heavy-duty tubing clamps offer a convenient and effective solution for flow control in bioprocessing applications. Designed for flexibility, these durable polysulfone clamps provide secure closure and accurate flow adjustment across multiple tubing materials and sizes. Their smooth contours facilitate easy cleaning, while the fully adjustable locking screw allows for quick installation and operation with just one hand. Manufactured in an ISO Class 8 cleanroom under strict GMP standards, these FDA-compliant clamps are suitable for high-purity fluid transfer applications such as sterile filling and media bag connections. They can be sterilized via autoclaving or gamma irradiation.
89,400.00 - ₹373,000.00INR / Pkg of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clamp Material Description
Availability Pricing
COMPARE
Polysulfone Heavy-Duty Tubing Clamps, 50/Pk
₹373,000.00INR / Pkg of 50
COMPARE
Polysulfone Heavy-Duty Tubing Clamps, 10/Pk
₹89,400.00INR / Pkg of 10