Heathrow Scientific® Adapt-A-Rack® Modular Tube Rack
Colorful multi-tube racks hold various tube sizes
  • Tubes stand upright regardless of tube diameter
  • Easy to view tube labels with open sides
  • It can safely withstand sterilization under pressure
Versatile rack design holds different-sized tubes from 12 to 30 mm dia in a secure, upright position. It offers a clear view of tube labels from any side for easy identification. The rack comes in fun colors to mix and match tops and bottoms for easy organization. Link racks together to increase capacity. Made of strong, autoclavable polyoxymethylene (POM) material.
4,700.00INR / Pkg of 2
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Max Tube Size (mm) Tubes Held
Availability Pricing
Heathrow Scientific Adapt-A-Rack® Modular Tube Rack System, blue & green, 7 to 50 mL tubes, 2/pk 0669616 Mfr # HS120184
COMPARE
Blue and green 30 4
₹4,700.00INR / Pkg of 2
Heathrow Scientific Adapt-A-Rack® Modular Tube Rack System, blue & yellow, 7 to 50 mL tubes, 2/pk 0669617 Mfr # HS120185
COMPARE
Blue and yellow 30 4
₹4,700.00INR / Pkg of 2
Heathrow Scientific Adapt-A-Rack® Modular Tube Rack System, pink & white, 7 to 50 mL tubes, 2/pk 0669618 Mfr # HS120186
COMPARE
Pink and white 30 4
₹4,700.00INR / Pkg of 2
Heathrow Scientific Adapt-A-Rack® Modular Tube Rack System, blue, 7 to 50 mL tubes, 2/pk 0669619 Mfr # HS120187
COMPARE
Blue 30 4
₹4,700.00INR / Pkg of 2
Heathrow Scientific Adapt-A-Rack® Modular Tube Rack System, white, 7 to 50 mL tubes, 2/pk 0669620 Mfr # HS120188
COMPARE
White 30 4
₹4,700.00INR / Pkg of 2
Heathrow Scientific Adapt-A-Rack® Modular Tube Rack System, pink, 7 to 50 mL tubes, 2/pk 0669621 Mfr # HS120189
COMPARE
Pink 30 4
₹4,700.00INR / Pkg of 2
Heathrow Scientific Adapt-A-Rack® Modular Tube Rack System, yellow, 7 to 50 mL tubes, 2/pk 0669622 Mfr # HS120190
COMPARE
Yellow 30 4
₹4,700.00INR / Pkg of 2
Heathrow Scientific Adapt-A-Rack® Modular Tube Rack System, green, 7 to 50 mL tubes, 2/pk 0669623 Mfr # HS120191
COMPARE
Green 30 4
₹4,700.00INR / Pkg of 2
Showing 1- 8 of 8