Hazard Chemical Bottle Pourers, PTFE
These worry-free polytetrafluoroethylene (PTFE) pourers have inert pouring surfaces — safely pour hazardous and corrosive chemicals. A breather vent allows direct and controlled pouring. Tethered strap keeps closure handy. Notch in tether keeps closure out of the way when pouring. A PTFE-encapsulated sealing ring ensures that bottle contents remain contaminant free. Fit standard reagent bottles; order bottles separately.
16,100.00 - ₹16,400.00INR / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap / Stopper Size Material
Availability Pricing
Dynalon Chemical Bottle Pourer, PTFE, 32-mm cap size WW-06038-02 Mfr # 310374-0002
COMPARE
GL32 Polytetrafluoroethylene (PTFE)
₹16,100.00INR / Pkg of 1
Dynalon Chemical Bottle Pourer, PTFE, 38-mm cap size WW-06038-04 Mfr # 310374-0004
COMPARE
GL38 Polytetrafluoroethylene (PTFE)
₹16,400.00INR / Pkg of 1
Dynalon Chemical Bottle Pourer, PTFE, 45-mm cap size WW-06038-06 Mfr # 310374-0001
COMPARE
GL45 Polytetrafluoroethylene (PTFE)
₹16,400.00INR / Pkg of 1