Hamilton® Syringe Needle with Kel-F Hub
Ideal for chromatography
  • Constructed of durable 304 stainless steel
  • Autoclavable
Hamilton® Syringe Needle with Kel-F hub (KF) can be used with Luer Tip (LT) and PTFE Luer Lock (TLL) syringes and connectors. Standard needle length is 2" / 51 mm.
16,800.00 - ₹37,900.00INR / Pkg of 6
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Needle Gauge Needle Tip Number Needle Tip Style
Availability Pricing
Hamilton Syringe Needle with Kel-F Hub, Beveled Tip, 22s G; 6/Pk 0793897 Mfr # 90138
COMPARE
22S 2 Sharp, beveled, curved, non-coring
₹21,500.00INR / Pkg of 6
Hamilton Syringe Needle with Kel-F Hub, Beveled Tip, 22 G; 6/Pk 0793898 Mfr # 90122
COMPARE
22 2 Sharp, beveled, curved, non-coring
₹16,800.00INR / Pkg of 6
Hamilton Syringe Needle with Kel-F Hub, Conical Tip, 26s G; 6/Pk 2570384 Mfr # 7746-12
COMPARE
26s 5 Conical
₹32,900.00INR / Pkg of 6
Hamilton Syringe Needle with Kel-F Hub, Conical Tip, 26 G; 6/Pk 2570385 Mfr # 7746-10
COMPARE
26 5 Conical
₹32,900.00INR / Pkg of 6
Hamilton Syringe Needle with Kel-F Hub, Conical Tip, 22s G; 6/Pk 2570386 Mfr # 7746-11
COMPARE
22s 5 Conical
₹37,900.00INR / Pkg of 6
Hamilton Syringe Needle with Kel-F Hub, Conical Tip, 22 G; 6/Pk 2570387 Mfr # 7746-06
COMPARE
22 5 Conical
₹32,900.00INR / Pkg of 6