Hamilton® Microliter Syringes for Agilent GC Autosamplers with Cemented Needle
Specifically designed for precision use in Agilent GC Autosamplers
  • Gas- and liquid-tight up to 2000 psi!
  • Syringes feature a 0.46 µL dead volume
  • AS point style is a special conical non-coring style needle point used on autosampler syringes for septum penetration
Hamilton® microliter syringes are designed for use with the Agilent GC Autosamplers 7693A, 7673, 7683, and 6850 ALS and feature a specially designed plunger button. Syringes include a cemented needle with AS needle point style, which is a special conical style needle point used on autosampler syringes. The non-coring needle point is recommended for septum penetration.
28,700.00 - ₹53,200.00INR / Pkg of 6
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Needle Gauge Needle Tip Style Syringe Capacity (µL)
Availability Pricing
Hamilton Microliter Low-Volume Syringe for Agilent GC Autosampler, Cemented Needle, Conical Tip, 5 µL, 23s G; 6/Pk WW-07938-28 Mfr # 87990
COMPARE
23s AS Style Conical Tip 5
₹53,200.00INR / Pkg of 6
Hamilton Microliter Low-Volume Syringe for Agilent GC Autosampler, Cemented Needle, Conical Tip, 5 µL, 23s-26s G; 6/Pk WW-07938-29 Mfr # 87994
COMPARE
23s-26s AS Style Conical Tip 5
₹53,200.00INR / Pkg of 6
Hamilton Microliter Low-Volume Syringe for Agilent GC Autosampler, Cemented Needle, Conical Tip, 10 µL, 23s G; 6/Pk WW-07939-30 Mfr # 80390
COMPARE
23s AS Style Conical Tip 10
₹39,400.00INR / Pkg of 6
Hamilton Microliter Low-Volume Syringe for Agilent GC Autosampler, Cemented Needle, Conical Tip, 10 µL, 23s-26s G; 1/Ea WW-07939-40 Mfr # 80391
COMPARE
23s-26s AS Style Conical Tip 10
₹28,700.00INR / Each