GW Instek PSW Series Multirange DC Power Supplies
Powerful stretch with multirange technology
  • Constant power output for multirange voltage and current operation
  • Constant voltage and current priority—ideal for battery and LED industries
  • Adjustable slew rate for added flexibility
  • Series and parallel configuration capabilities
  • High efficiency and high power density
  • USB, LAN, and analog control standard interfaces, with optional USB-GPIB adapter
443,000.00 - ₹599,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek PSW 30-60 DC Power Supply, 30 V, 36 A 2005011 Mfr # PSW 30-36
COMPARE
DC Power Supply, 30 V, 36 A
₹466,000.00INR / Each
GW Instek PSW 80-13.5 DC Power Supply, 80 V, 13.5 A 2005014 Mfr # PSW 80-13.5
COMPARE
DC Power Supply, 80 V, 13.5 A
₹443,000.00INR / Each
GW Instek PSW 80-27 DC Power Supply, 80 V, 27 A 2005015 Mfr # PSW 80-27
COMPARE
DC Power Supply, 80 V, 27 A
₹570,000.00INR / Each
GW Instek PSW 160-7.2 DC Power Supply, 160 V, 7.2 A 2005017 Mfr # PSW 160-7.2
COMPARE
DC Power Supply, 160 V, 7.2 A
₹599,000.00INR / Each
GW Instek PSW 800-1.44 DC Power Supply, 800 V, 1.44 A 2005023 Mfr # PSW 800-1.44
COMPARE
DC Power Supply, 800 V, 1.44 A
₹579,000.00INR / Each