GVS Dilution Bottles
Eliminate the time and money spent on preparing your own dilution vials
  • Prefilled bottles make sample preparation easy and consistent
  • Final pH for all solutions is 7.2 pH ±0.2 pH at 25°C
  • Sterility makes these suitable for sensitive microbiological and pharmaceutical processes
Ensure consistent dilutions and reduce the risk of contamination with these prefilled, sterile dilution bottles. Designed with an easy open, flip-top, plastic container with tamper-evident seal to produce an accurate and safe sample dilution. These buffers are ideal for sample dilution of water, dairy products, foods and pharmaceuticals prior to microbiological testing. Choose from Butterfield’s Phosphate Buffer or Phosphate Buffer with Magnesium Chloride.
Butterfield’s Buffer contains monobasic potassium phosphate and is used extensively in the food, dairy, and pharmaceutical industries to ensure the stable pH during the testing process and does not interfere with microbial growth. Offered in 90 ml and 99 ml volumes for easy 1:10 and 1:100 dilutions. Butterfield’s buffer is recommended as a general diluent in laboratory procedures by the Federal Drug Administrations and in the Bacteriological Analytical Manual. It is recommended by APHA for the recovery of injured microorganisms from dairy and food samples.
Phosphate Buffer with magnesium chloride is used as the diluents for the preparation of dilutions in plate counts in the dairy and food industries. This product is prepared according to Standard Methods for the Examination of Water and Wastewater for use in water testing. It Contains deionized water, monopotassium phosphate, and magnesium chloride.
34,700.00 - ₹66,300.00INR / Pkg of 72
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Element/Ion Volume (mL) Description
Availability Pricing
GVS Dilution Bottle, Butterfield's Buffer, 99 mL; 72/PK 3615317 Mfr # 10498503S
COMPARE
Butterfield's Buffer 99 Dilution Bottle, Butterfield's Buffer, 99 mL; 72/PK
₹66,300.00INR / Pkg of 72
GVS Dilution Bottle, Phosphate Buffer with Magnesium Chloride, 99 mL; 72/PK 3615319 Mfr # 10498505
COMPARE
Phosphate Buffer with Magnesium Chloride 99 Dilution Bottle, Phosphate Buffer with Magnesium Chloride, 99 mL; 72/PK
₹34,700.00INR / Pkg of 72