Glass/Stainless Steel Desiccators
Glass door and sides let you easily view contents
High-polish 302 stainless steel cabinets have 3/16" thick glass door and sides for observing contents. Door has double-action lock, one-piece rubber gasket, and full-length hinge. Manual pressure vent valve lets you release pressure. Choose from flexboard or SS shelves; shelves measure 9-3/4"L x 8"W and have twelve 1/2" height adjustments. SS desiccant tray holds one pound of desiccant. Desiccator measures 12-1/2"W x 12-1/4"H x 12-1/2"D overall with an 11-1/2"W x 12"H x 10"D interior.
289,000.00 - ₹298,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Exterior Depth (in) Exterior Height (in) Exterior Width (in)
Availability Pricing
Boekel Scientific 1340 Glass/SS Desiccator w/Two Flexboard Shelves; 12.5"W x 12.25"H x 12.5"D WW-08901-50 Mfr # 1340
COMPARE
12.5 12.25 12.5
₹289,000.00INR / Each
Boekel Scientific 1342 Glass/SS Desiccator w/Two SS Shelves; 12.5"W x 12.25"H x 12.5"D WW-08901-60 Mfr # 1342
COMPARE
12.5 12.25 12.5
₹298,000.00INR / Each