GHS Flame Pictogram Labels
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Flame pictogram indicates the following potential hazards: flammables, pyrophorics, self-heating, emits flammable gas, self-reactives, and organic peroxides.
6,500.00 - ₹16,300.00INR / Roll of 250
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Quantity
Availability Pricing
Accuform LZH601PS2 GHS Label, Flame Pictogram, 1" x 1", adhesive coated paper, 250/roll 5071248 Mfr # LZH601PS2
COMPARE
1 Adhesive Paper 250/roll
₹6,500.00INR / Roll of 250
Accuform LZH601PS5 GHS Label, Flame Pictogram, 1" x 1", adhesive coated paper, 500/roll 5071249 Mfr # LZH601PS5
COMPARE
1 Adhesive Paper 500/roll
₹10,700.00INR / Roll of 500
Accuform LZH601EV2 GHS Label, Flame Pictogram, 1" x 1", adhesive poly sheet, 250/roll 5071250 Mfr # LZH601EV2
COMPARE
1 Adhesive Poly 250/roll
₹10,200.00INR / Roll of 250
Accuform LZH601EV5 GHS Label, Flame Pictogram, 1" x 1", adhesive poly sheet, 500/roll 5071251 Mfr # LZH601EV5
COMPARE
1 Adhesive Poly 500/roll
₹16,300.00INR / Roll of 500
Accuform LZH611PS2 GHS Label, Flame Pictogram, 2" x 2", adhesive coated paper, 250/roll 5071252 Mfr # LZH611PS2
COMPARE
2 Adhesive Paper 250/roll
₹7,800.00INR / Roll of 250
Accuform LZH611PS5 GHS Label, Flame Pictogram, 2" x 2", adhesive coated paper, 500/roll 5071253 Mfr # LZH611PS5
COMPARE
2 Adhesive Paper 500/roll
₹13,200.00INR / Roll of 500