GHS Flame Over Circle Pictogram Labels
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Flame Over Circle pictogram indicates the following potential hazard: oxidizers
6,500.00 - ₹16,300.00INR / Roll of 250
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Quantity
Availability Pricing
Accuform LZH602PS2 GHS Label,Flame Over Circle Pictogram,1" x 1",adhesive paper,250/roll 5071256 Mfr # LZH602PS2
COMPARE
1 Adhesive Paper 250/roll
₹6,500.00INR / Roll of 250
Accuform LZH602EV2 GHS Label,Flame Over Circle Pictogram,1" x 1",adhesive poly,250/roll 5071258 Mfr # LZH602EV2
COMPARE
1 Adhesive Poly 250/roll
₹10,200.00INR / Roll of 250
Accuform LZH602EV5 GHS Label,Flame Over Circle Pictogram,1" x 1",adhesive poly,500/roll 5071259 Mfr # LZH602EV5
COMPARE
1 Adhesive Poly 500/roll
₹16,300.00INR / Roll of 500