GHS Exclamation Point Pictogram Labels
GHS (Globally Harmonized System) is a worldwide program for classifying chemicals in the workplace. GHS recognizes nine hazard pictograms. The pictograms identify the hazard which helps users take proper precautions to minimize adverse effects.
GHS Exclamation Point pictogram indicates the following potential hazard: irritant (skin and eye), skin sensitizer, acute toxicity, narcotic effects, respiratory tract irritant, hazardous to ozone layer.
4,600.00 - ₹11,600.00INR / Roll of 500
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Material Quantity
Availability Pricing
Accuform LZH607PS2 GHS Label,Exclamation Point Pictogram,1" x 1",adhesive paper,250/roll WW-50713-00 Mfr # LZH607PS2
COMPARE
1 Adhesive Paper 250/roll
₹4,600.00INR / Roll of 250
Accuform LZH607PS5 GHS Label,Exclamation Point Pictogram,1" x 1",adhesive paper,500/roll WW-50713-01 Mfr # LZH607PS5
COMPARE
1 Adhesive Paper 500/roll
₹6,200.00INR / Roll of 500
Accuform LZH607EV2 GHS Label,Exclamation Point Pictogram,1" x 1",adhesive poly,250/roll WW-50713-02 Mfr # LZH607EV2
COMPARE
1 Adhesive Poly 250/roll
₹6,700.00INR / Roll of 250
Accuform LZH607EV5 GHS Label,Exclamation Point Pictogram,1" x 1",adhesive poly,500/roll WW-50713-03 Mfr # LZH607EV5
COMPARE
1 Adhesive Poly 500/roll
₹9,700.00INR / Roll of 500
Accuform LZH617PS2 GHS Label,Exclamation Point Pictogram,2" x 2",adhesive paper,250/roll WW-50713-04 Mfr # LZH617PS2
COMPARE
2 Adhesive Paper 250/roll
₹4,800.00INR / Roll of 250
Accuform LZH617PS5 GHS Label,Exclamation Point Pictogram,2" x 2",adhesive paper,500/roll WW-50713-05 Mfr # LZH617PS5
COMPARE
2 Adhesive Paper 500/roll
₹8,100.00INR / Roll of 500
Accuform LZH617EV2 GHS Label,Exclamation Point Pictogram,2" x 2",adhesive poly,250/roll WW-50713-06 Mfr # LZH617EV2
COMPARE
2 Adhesive Poly 250/roll
₹7,400.00INR / Roll of 250
Accuform LZH617EV5 GHS Label,Exclamation Point Pictogram,2" x 2",adhesive poly,500/roll WW-50713-07 Mfr # LZH617EV5
COMPARE
2 Adhesive Poly 500/roll
₹11,600.00INR / Roll of 500
Showing 1- 8 of 8