Gay-Lussac Bottles
Conveniently determine the density of liquid samples
  • Choose from unadjusted and adjusted models
  • Unadjusted models must be calibrated with water before use
  • Adjusted bottles are calibrated to ASTM Test Reference D-369 - just fill with sample and weigh
Gay-Lussac bottles have ground-in perforated glass stoppers with outer ground-on cap to reduce evaporation.
36,700.00 - ₹74,400.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Outside Diameter (OD) (in)
Availability Pricing
COMPARE
10 Glass 1 7/32
₹36,700.00INR / Each
COMPARE
2 Glass 43/64
₹65,100.00INR / Each
COMPARE
5 Glass 3/4
₹65,900.00INR / Each
COMPARE
10 Glass 1 7/32
₹66,600.00INR / Each
COMPARE
25 Glass 1 17/32
₹71,200.00INR / Each
COMPARE
50 Glass 1 57/64
₹73,000.00INR / Each
COMPARE
100 Glass 2 9/32
₹74,400.00INR / Each
Showing 1- 7 of 7