Fomblin Vacuum Pump Oil

Formulated to ensure optimum pump performance
  • Compatible with corrosive gases, oxygen and hydrogen, plastics, elastomers, and metals
  • Increases the mean-time between failure
  • Ideal for use in chemical processes
The YL-Vac PFPE vacuum pump oils are formulated to ensure optimum pump performance and pump life They are ideal for use in chemical processes which require inert mechanical vacuum pump oils.
94,000.00INR / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Specific Gravity Vapor Pressure (torr) Volume (mL)
Availability Pricing
Fomblin Y L-VAC 06/6 PFPE Pump Inert Oil, 1 kg WW-79751-70 Mfr # 06/6 1 KG
COMPARE
1.88 8x10-7 517
₹94,000.00INR / Each
Fomblin Y L-VAC 16/6 PFPE Pump Inert Oil, 1 kg WW-79751-71 Mfr # 16/6,1 KG
COMPARE
1.89 2x10-6 517
₹94,000.00INR / Each
Fomblin Y L-VAC 25/6 PFPE Pump Inert Oil, 1 kg WW-79751-72 Mfr # 25/6,1 KG
COMPARE
1.9 6x10-8 517
₹94,000.00INR / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat