Fluke SMFT-1000 Multifunction PV Analyzer Kits
All-in-one test solution to verify solar system performance and safety while expediting client reporting
  • Meets IEC 62446-1 standards for Category 1 and Category 2 tests
  • Open-circuit voltage (VOC) measurement at the PV module/string up to 1000 V DC
  • Short-circuit (ISC) current measurement at the PV module/string up to 20 A DC
  • On-location I-V curve results compares manufacturer I-V curve data to measured data on the analyzer screen
  • Compatible with Fluke TruTest™ data management software
Test that PV systems are performing to their optimal power output as well as operating safely with the Fluke SMFT-1000 multifunction testers with I-V curve tracing. Designed for PV professionals that provide installation, commissioning and maintenance services to systems that operate at 1000 V DC or under, the SMFT-1000 kits provide a complete PV testing solution that conforms to IEC 62446-1 standards. Through Fluke’s TruTest™ Software, measurement data from solar site installation and commissioning testing can be easily imported, organized, and analyzed for effortless reporting without having to bring a laptop on-site.
To perform precise I-V curve measurements, real time irradiance and temperature data is needed. The included IRR2-BT irradiance meter wirelessly connects to the SMFT-1000 to communicate data in real time, providing the most accurate I-V curve measurements possible. If the wireless connection is interrupted for any reason, the IRR2-BT will continue recording data for up to 17 hours that can later be matched to tests taken with the SMFT-1000.
1,919,000.00 - ₹1,933,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max AC Voltage (Voltage) Min AC Voltage (Voltage) Description
Availability Pricing
Fluke Solar Tools Kit with Multifunction PV Analyzer and Solar Irradiance Meter 2005130 Mfr # FLK-SMFT-1000/KIT
COMPARE
700 5 Solar Tools Kit with Multifunction PV Analyzer and Solar Irradiance Meter
₹1,919,000.00INR / Each
Fluke Solar Tools Pro Kit with Multifunction PV Analyzer, Solar Irradiance Meter, TruTest™ Software, and MC4 Leads 2005131 Mfr # FLK-SMFT-1000/PRO
COMPARE
700 5 Solar Tools Pro Kit with Multifunction PV Analyzer, Solar Irradiance Meter, TruTest™ Software, and MC4 Leads
₹1,933,000.00INR / Each