Fluke Precision Digital Test Gauges
Portable, high-quality gauge allows fast and accurate calibration test results
  • Precision pressure measurement from 15 psi/1 bar to 10,000 psi/690 bar
  • Accuracy to ±0.05% of full scale
  • Easy-to-use interface with no difficult menus
  • Rugged construction for reliable performance
  • CSA Class 1, Div 2, Groups A-D rating
  • ATEX rating II 3 G Ex nA IIB T6
  • Log up to 8493 pressure measurements to memory using 700G/TRACK software
The 700G series of precision test gauges are accurate to ±0.05% full-scale — ideal for use as a calibration reference. The four-button display allows you to zero, set min/max, set 23 units of measure, as well as many other functions. Using the 700G/TRACK software with the 700G series gauges lets you perform real-time data logging to a PC, download logging configurations to the gauges for a remote logging event, and configure logging event reading rate, duration and measurement units. Software also uploads measurements logged remotely and displays or export measurements. Software includes communication cable for connection to a PC.
Combine these Fluke 700G gauges with the 700PTPK or 700HTPK pump kits for a complete pressure testing solution for up to 600 psi with the PTP-1 pneumatic pump and up to 10,000 psi with the HTP-2 hydraulic pump.
544,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Fluke 700G10 Digital Test Gauge 2000 PSI 1/4 NPT(M) 6808804 Mfr # FLUKE-700G10
COMPARE
±0.05% full-scale (positive pressure); ±0.1% full-scale (negative pressure) 2000 0.1 psi
₹544,000.00INR / Each
Fluke 700GA4 Digital Test Gauge 15 PSIA1/4 NPT(M) 6808806 Mfr # FLUKE-700GA4
COMPARE
±0.05% full-scale 29.7 0.001 psi
₹544,000.00INR / Each