Fluke Networks MicroScanner™ Industrial Ethernet Cable Verifier Kits
Perform continuity tests on RJ45, M12X, M12D, and M8D terminated cables
  • Displays cable length, wiremap and distance to fault on a single screen
  • Identifies remote ID locators and detects Ethernet ports
  • Generates IntelliTone™ and normal analog toning signals
Quickly determine the location of opens, shorts, and crosswires with the MicroScanner industrial Ethernet cable verifier kits. Since cable failures result in costly production downtime, it is vital to replace or repair broken cables fast. View cable length, wiremap and distance to fault on a large LCD, making it easy to identify the location of the fault. The MicroScanner PoE models help determine if a problem is caused by insufficient power or the powered device.
Select from three test modes: cable test, tone, or power over Ethernet (PoE). Use the cable test mode to measure the cable length, verify wiremap, identify remote ID locators, and detect Ethernet ports. Tone mode helps locate cables by generating IntelliTone™ and normal analog signals. In the PoE mode, MicroScanner 2 verifier solicits and detects the presence of 802.3af compatiblePoE devices while Microscanner PoE verifiers solicits and detects the presence of 802.3af, at, bt, and UPOE (Cisco’s Universal Power over Ethernet) compatible PoE devices.
698,000.00 - ₹876,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Function Range Description
Availability Pricing
Fluke Networks MicroScanner™ PoE Industrial Ethernet Cable Verifier Kit 1778474 Mfr # MS-POE-IE
COMPARE
Cable test, tone, and PoE 460 m (1500 ft) MicroScanner™ PoE Industrial Ethernet Cable Verifier Kit
₹769,000.00INR / Each
Fluke Networks MicroScanner™ PoE Industrial Ethernet Cable Verifier Kit with Intellitone™ Pro 200 Probe 1778475 Mfr # MS-POE-IE-KIT
COMPARE
Cable test, tone, and PoE 460 m (1500 ft) MicroScanner™ PoE Industrial Ethernet Cable Verifier Kit with Intellitone™ Pro 200 Probe
₹876,000.00INR / Each
Fluke Networks MicroScanner™ 2 Industrial Ethernet Cable Verifier Kit 1778476 Mfr # MS2-100-IE
COMPARE
Cable test, tone, and PoE 460 m (1500 ft) MicroScanner™ 2 Industrial Ethernet Cable Verifier Kit
₹698,000.00INR / Each