Fluke ii915 Precision Acoustic Imager
An easy way to detect, locate and analyze partial discharge in one tool
  • MecQ™ mechanical scanning can detect bad bearings in conveyor systems before they worsen
  • Quick, simple partial discharge detection and PD testing
  • Reduce outages and increase uptime
  • Cut costs and save energy by finding and fixing PD
  • PDQ Mode to capture and analyze partial discharge
The ii915 Precision Acoustic Imager is engineered to locate partial discharge, corona discharge, as well as gas and vacuum leaks. Partial discharge that goes unchecked can cause blackouts, fires, explosions, or death from arc flashes. Quickly scan the area or environment and get all the pictures or videos you need with the simple one button capture and large 7” touchscreen LCD. Without cumbersome secondary cables or devices, you have full mobility and greater flexibility to swap between detection of the leak/PD and fixing the leak/PD. With SoundSight™ technology, the ii915 translates the sounds that it hears into a visual representation so that you can quickly locate problem areas. The higher frequency capability of the ii915 allows for earlier detection to facilitate maintenance planning. The imager also features 32 GB memory, Wi-Fi, tripod mount, and replaceable batteries.
9,814,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Fluke ii915 Precision Acoustic Imager 3974960 Mfr # FLUKE-ii915
COMPARE
Precision Acoustic Imager
₹9,814,000.00INR / Each