Fluke® 393 FC CAT III 1500 V True-rms Clamp Meter with iFlex
Measure safely with this CAT III 1500 V rated clamp meter
  • Thin jaw for access to cables in crowded combiner boxes
  • Sturdy IP54 rated for outdoor use
  • DC power measurement, showing readings in kVA
  • Audio Polarity indicator helps prevent accidental miswires
  • Visual Continuity turns provide a bright green light in the display, ideal when working in dark and noisy environments
  • Logging and reporting of test results via Fluke Connect software
  • When measuring AC current the included iFlex flexible current probe gives you unparalleled access to cable is tight spaces
The 393 FC CAT III 1500 V True-rms Clamp Meter with iFlex is designed for technicians who work in high voltage DC environment: solar arrays, wind power, electric railways, data centers battery banks for uninterruptible power supplies. The clamp will measure up to 1500 V DC, 1000 V AC, and up to 999.9 A DC or AC through the clamp jaw. The included iFlex flexible current probe extended AC current measurements up to 2500 amps. This clamp has a thin jaw, giving you access to cables in crowded combiner boxes. Test leads are designed with your work in mind, and are also rated to CAT III 1500 V DC.
Bundles are also available, combining the clamp meter with a wide array of solar tools to easily meet your needs.
244,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) DC Max (Volts) Description
Availability Pricing
Fluke 393 FC CAT III 1500 V True-rms Solar Clamp Meter with iFlex 2003740 Mfr # FLUKE-393 FC
COMPARE
1000 1500 CAT III 1500 V True-rms Solar Clamp Meter with iFlex
₹244,000.00INR / Each