Fluke 120B Series ScopeMeter® Handheld Oscilloscopes
Maximize uptime and easily collaborate to make confident decisions
  • Fluke Connect® allows access to data remotely via smart phones and tablets
  • Advanced triggering technology provides a stable, reliable, and repeatable test
  • Automatically displays appropriate parameters based on waveform qualities
  • Troubleshoot and analyze with three testing instruments in one—oscilloscope, multimeter, and recorder
This handheld digital oscilloscope features advanced connectivity options to make data analysis, data sharing, and maintenance or troubleshooting decisions faster and easier. Included WiFi adapter communicates with PCs and mobile devices on wireless LAN using the downloadable Fluke Connect app. Optional optically isolated USB interface cable allows direct connection to PC where Wi-Fi is unavailable (order separately). Choose a ScopeMeter kit for FlukeView® software, which enables further collaboration, data analysis, and archiving of critical test information.
Operation is simplified by two programs to give you quick access to the data you need. Connect-and-View™ hands-free triggering technology recognizes signal patterns, sets up the correct trigger program, and automatically captures waveforms. IntellaSet™ uses proprietary algorithms to analyze measured waveforms, then displays only the pertinent numerical data for that wave type.
Available for sale in the United States and Canada only.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bandwidth (MHz) Sample Rate (MS/s) Description
Availability Pricing
Fluke 123B ScopeMeter Industrial Handheld Oscilloscope, 20 MHz, N America 2000002 Mfr # FLUKE-123B/NA
COMPARE
20 40 ScopeMeter Industrial Handheld Oscilloscope, 20 MHz, N America
Call For Price
Fluke 124B ScopeMeter Industrial Handheld Oscilloscope, 40 MHz, N America 2000003 Mfr # FLUKE-124B/NA
COMPARE
40 40 ScopeMeter Industrial Handheld Oscilloscope, 40 MHz, N America
Call For Price
Fluke 125B/S ScopeMeter Industrial Handheld Oscilloscope Kit, 40 MHz, N America 2000012 Mfr # FLUKE-125B/NA/S
COMPARE
40 40 ScopeMeter Industrial Handheld Oscilloscope Kit, 40 MHz, N America