Floating Microtube Racks, polymethylpentene
Incubate 8 to 20 microtubes in a beaker of water
The perfect rack for use in microbiology and immunology labs. Made especially for use with beakers, the space-saving round bubble racks are ideal for small quantities of tubes. The racks are filled from the top and tubes are easily removed by pressing the rack down on the bench until the legs touch, releasing the tubes. Molded of buoyant polymethylpentene with tiny bubbles throughout for additional buoyancy. Tube holes are arranged in a circular pattern.
3,200.00 - ₹3,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubes Held Description
Availability Pricing
Scienceware F18875-0400 Floating Microcentrifuge Tube Rack (holds 8 X 1.5 mL Microtubes) WW-06274-12 Mfr # F18875-0400
COMPARE
8 Floating Microcentrifuge Tube Rack (holds 8 X 1.5 mL Microtubes)
₹3,200.00INR / Each
Scienceware F18875-1000 Floating Microcentrifuge Tube Rack (holds 20 X 1.5 mL Microtubes) WW-06274-13 Mfr # F18875-1000
COMPARE
20 Floating Microcentrifuge Tube Rack (holds 20 X 1.5 mL Microtubes)
₹3,600.00INR / Each
Scienceware F18875-1600 Floating Microcentrifuge Tube Rack (holds 20 X 0.5 mL Tubes) WW-06274-14 Mfr # F18875-1600
COMPARE
20 Floating Microcentrifuge Tube Rack (holds 20 X 0.5 mL Tubes)
₹3,800.00INR / Each