FLIR SV88™ and SV89™ Vibration Monitoring Kits
FLIR SV88™ and SV89™ Vibration Monitoring Kits offer advanced vibration analytics and rugged wireless reliability - ensuring your critical equipment stays running, and your downtime stays at zero
  • Get real-time analysis of GRMS, VRMS, FFT, crest factor, kurtosis, and other key vibration metrics
  • Install with ease in various environments thanks to wireless connectivity and a 50-meter range
  • Rely on rugged, IP66-rated construction for consistent performance in harsh industrial conditions
  • Integrate effortlessly with your existing systems using Modbus, MQTT, and OPC UA protocols
Whether you're managing a plant floor or overseeing rotating equipment across multiple facilities, the FLIR SV88 and SV89 are your go-to sensors for predictive maintenance. With configurable data capture rates and high-frequency analysis up to 10 kHz, these compact sensors detect even the slightest signs of misalignment, bearing wear, or gear issues before they turn into failures.
Each sensor communicates wirelessly with the included GW66 Gateway, enabling real-time data transmission and centralized monitoring. Combined with robust environmental protection and a 3-year warranty, FLIR’s vibration monitoring solution gives you peace of mind—and the power to act fast. Say goodbye to unplanned downtime and hello to smarter, data-driven maintenance.
379,000.00 - ₹429,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem Availability Pricing
Flir SV88™ Vibration Monitoring Solution Kit 3975937 Mfr # SV88-KIT
COMPARE
₹379,000.00INR / Each
Flir SV89™ Vibration Monitoring Solution Kit 3975938 Mfr # SV89-KIT
COMPARE
₹429,000.00INR / Each