FLIR® Exx-Series Thermal Cameras
Features onboard inspection routing to speed up surveys and simplifying reporting
  • Improved resolution up to 640 x 480 pixels
  • Vibrant, 4” optically-bonded LCD touch screen with intuitive new user interface
  • Includes Multi-Spectral Dynamic Imaging System (MSX®) to add visible spectrum definition to thermal images in real time
  • Rugged design is IP54 rated for demanding applications
  • Connect to mobile devices using Wi-Fi or Bluetooth®
  • Offers MeterLink™ to transfer, via Bluetooth™, the data acquired by a FLIR measurement instrument into the infrared camera and implement it on the infrared image for coordinated documentation
Each FLIR Exx-Series camera is packed with features to find hot spots, detect early signs of building deficiencies, troubleshoot electrical and mechanical systems, and prevent problems before they cause serious damage. The Exx-series is the first to include an inspection route camera option automatically enabled. Designed for thermographers, the inspection route option guides a user along a predefined route of points to collect images and data in a structured fashion. Users can build their route plan in the Thermal Studio Pro software. There is no limit on the number of inspection points that can be added to a route.
The EST-models include elevated skin temperature measurement in addition to the basic features. EST™ thermal screening solution detects and visualizes heat to quickly identify individuals with an elevated skin temperature. The on-camera FLIR Screen-EST Mode offers visual positioning guides, automatic sampling to maintain an updated temperature average, and graphic pass/fail indicators for results, reducing the burden on screening station operators. Compatibility with FLIR Screen-EST Desktop software, an integrated tripod mount, and external power makes this camera a good fit for permanent installations.
2,209,000.00 - ₹3,604,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Pixels / Thermal Resolution Description
Availability Pricing
Flir E54 Thermal Imaging Camera with MSX®, 320 x 240 Resolution (76,800 Pixels) and 24° Lens, with Starter Thermal Dataset 3975824 Mfr # 79302-0201
COMPARE
650 320 × 240 pixels E54 Thermal Imaging Camera with MSX®, 320 x 240 Resolution (76,800 Pixels) and 24° Lens, with Starter Thermal Dataset
₹2,209,000.00INR / Each
Flir E76 Thermal Imaging Camera with MSX®, 320 x 240 Resolution (76,800 Pixels) and 24° Lens, with Starter Thermal Dataset 3975826 Mfr # Flir E76-24
COMPARE
1000 320 × 240 pixels E76 Thermal Imaging Camera with MSX®, 320 x 240 Resolution (76,800 Pixels) and 24° Lens, with Starter Thermal Dataset
₹2,958,000.00INR / Each
Flir E76 Thermal Imaging Camera with MSX®, 320 x 240 Resolution (76,800 Pixels) and 42° Lens, with Starter Thermal Dataset 3975827 Mfr # Flir E76-42
COMPARE
1500 464 × 348 pixels E76 Thermal Imaging Camera with MSX®, 320 x 240 Resolution (76,800 Pixels) and 42° Lens, with Starter Thermal Dataset
₹2,958,000.00INR / Each
Flir E76 Thermal Imaging Camera with MSX®, 320 x 240 Resolution (76,800 Pixels) and 42° + 14° Lens, with Starter Thermal Dataset 3975831 Mfr # Flir E76-42-14
COMPARE
1000 320 × 240 pixels E76 Thermal Imaging Camera with MSX®, 320 x 240 Resolution (76,800 Pixels) and 42° + 14° Lens, with Starter Thermal Dataset
₹3,604,000.00INR / Each
Flir E86 Thermal Imaging Camera with MSX®, 464 x 348 Resolution (161,472 Pixels) and 24° Lens, with Starter Thermal Dataset 3975833 Mfr # Flir E86-24
COMPARE
1500 464 × 348 pixels E86 Thermal Imaging Camera with MSX®, 464 x 348 Resolution (161,472 Pixels) and 24° Lens, with Starter Thermal Dataset
Call For Price
Flir E86 Thermal Imaging Camera with MSX®, 464 x 348 Resolution (161,472 Pixels) and 24° + 42° Lens, with Starter Thermal Dataset 3975835 Mfr # Flir E86-24-42
COMPARE
1500 464 × 348 pixels E86 Thermal Imaging Camera with MSX®, 464 x 348 Resolution (161,472 Pixels) and 24° + 42° Lens, with Starter Thermal Dataset
Call For Price
Flir E96 Thermal Imaging Camera with MSX®, 640 x 480 Resolution (307,200 Pixels) and 24° Lens, with Starter Thermal Dataset 3975840 Mfr # Flir E96-24
COMPARE
1500 640 × 480 pixels E96 Thermal Imaging Camera with MSX®, 640 x 480 Resolution (307,200 Pixels) and 24° Lens, with Starter Thermal Dataset
Call For Price
Flir E96 Thermal Imaging Camera with MSX®, 640 x 480 Resolution (307,200 Pixels) and 24° + 42° Lens, with Starter Thermal Dataset 3975842 Mfr # Flir E96-24-42
COMPARE
1500 640 × 480 pixels E96 Thermal Imaging Camera with MSX®, 640 x 480 Resolution (307,200 Pixels) and 24° + 42° Lens, with Starter Thermal Dataset
Call For Price
Showing 1- 8 of 8