Flammable Cabinets
Reduce fire risks and protect workers
  • Store chemicals safely in the workplace
  • Enhance fire safety through door-lock system, and fireproof and explosion-proof vents
  • FM and CE certifications in strict compliance with FM approval standard for storage cabinets
Safety cabinets for storing flammable liquids (low and median flash
point liquids, flash point <37.8).
Safety storage cabinets feature 18-gauge steel, all-welded, double fireproof steel panel, insulating air barrier
construction for protection and durability. Fireproofing vents at both sides of the cabinet (one on the top and
another on the bottom) effectively reduce the concentration of volatile substances and prevent fire hazards.
Reinforced shelves are adjustable and feature built-in troughs and leakproof wells to contain spills. The
manually-closing doors can be fully opened to 180° for easy access. Three-point linkage latch can be
padlocked to ensure complete closure for better security.
In storing chemicals, cabinet colors and warning labels are generally used to identify different chemicals, so if
a fire occurs, it is easy for firemen to quickly locate hazardous chemicals.
50,800.00 - ₹352,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Number of Shelves
Availability Pricing
SYSBEL WA810100 10 Gal/38L,1 door, manual WW-15976-67 Mfr # WA810100
COMPARE
38 1
₹67,200.00INR / Each
SYSBEL WA810120 12 Gal/45L,1 door, manual WW-15976-68 Mfr # WA810120
COMPARE
45 1
₹70,500.00INR / Each
SYSBEL WA810220 22 Gal/83L,1 door, manual WW-15976-69 Mfr # WA810220
COMPARE
83 3
₹119,000.00INR / Each
SYSBEL WA810300 30 Gal/114L,2 door, manual WW-15976-70 Mfr # WA810300
COMPARE
114 1
₹107,000.00INR / Each
SYSBEL WA810450 45 Gal/170L,2 door, manual WW-15976-71 Mfr # WA810450
COMPARE
170 2
₹134,000.00INR / Each
SYSBEL WA810540 54 Gal/204L,1 door, manual WW-15976-72 Mfr # WA810540
COMPARE
204 3
₹210,000.00INR / Each
SYSBEL WA810600 60 Gal/227L,2 door, manual WW-15976-73 Mfr # WA810600
COMPARE
227 1
₹150,000.00INR / Each
SYSBEL WA810860 90 Gal/340L,2 door, manual WW-15976-74 Mfr # WA810860
COMPARE
340 2
₹162,000.00INR / Each
SYSBEL WA810550 55 Gal/207L,2 door, manual WW-15976-75 Mfr # WA810550
COMPARE
207 2
₹220,000.00INR / Each
SYSBEL WA811100 110 Gal/415L,2 door, manual WW-15976-76 Mfr # WA811100
COMPARE
415 1
₹336,000.00INR / Each
SYSBEL WA810115 115 Gal/434L,2 door, manual WW-15976-77 Mfr # WA810115
COMPARE
434 3
₹352,000.00INR / Each
SYSBEL WA810040 Flammable Cabinet, 15 L; 1 door WW-16017-22 Mfr # WA810040
COMPARE
15 1
₹50,800.00INR / Each
Showing 1- 12 of 12