Flame-Resistant Lab Coats
Flame-resistant lab coats protect you when working with flammable chemicals
Constructed of 100% cotton, lab coats feature three outside pockets, side vent openings, hemmed sleeve ends, and a lapel collar. Rated to Arc Thermal Protective Value (ATPV) of 7.7 calories/cm2 and a Hazard Risk Category (HRC) of HRC1.
16,700.00 - ₹25,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Length (in) Size
Availability Pricing
VF Workwear KEL2LB-SMALL Flame-Resistant Lab Coat, Light Blue, small WW-86468-30 Mfr # KEL2LB-SMALL (PACK BY PERSON)
COMPARE
Blue 41.5 Small
₹16,700.00INR / Each
VF Workwear KEL2LB-MEDIUM Flame-Resistant Lab Coat, Light Blue, medium WW-86468-31 Mfr # KEL2LB-MEDIUM (PACK BY PERSON)
COMPARE
Blue 41.5 Medium
₹16,700.00INR / Each
VF Workwear KEL2LB-LARGE Flame-Resistant Lab Coat, Light Blue, large WW-86468-32 Mfr # KEL2LB-LARGE (PACK BY PERSON)
COMPARE
Blue 41.5 Large
₹25,000.00INR / Each
VF Workwear KEL2LB-ARGE Flame-Resistant Lab Coat, Light Blue, extra large WW-86468-33 Mfr # KEL2LB-XLARGE (PACK BY PERSON)
COMPARE
Blue 41.5 X-Large
₹25,000.00INR / Each