Falcon® Polystyrene Centrifuge Tubes, Conical Bottom
Polystyrene centrifuge tubes with secure dome seal caps for reliable sample handling
  • Dome seal screw caps provide a secure, full-contact closure with positive seal over the full circumference to help prevent leaks during handling and transport
  • Designed for low-speed centrifugation up to ~1,800 × g
  • Available in both bulk and racked formats to support different workflow needs
  • Clear tube design with dark blue graduations and marking areas supports easy sample identification
These Falcon® 15 mL centrifuge tubes are designed for routine laboratory workflows where secure sample handling is essential. Featuring a dome seal screw cap, they provide a reliable, full-circumference seal that helps reduce the risk of leakage during transport and processing. Suitable for centrifugation up to approximately 1,800 × g, these tubes are ideal for low- to moderate-speed applications. Available in both bulk and racked formats, they offer flexibility for different lab environments while maintaining consistent performance and ease of use.
8,400.00 - ₹76,000.00INR / Case of 500
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Packaging Tube Capacity (mL)
Availability Pricing
Falcon 15 mL Polystyrene Centrifuge Tube, Conical Bottom, with Dome Seal Screw Cap, Sterile, 50/Bag, 500/Case 4846028 Mfr # 352095
COMPARE
50/Bag, 500/Case 15
₹71,800.00INR / Case of 500
Falcon 15 mL Polystyrene Centrifuge Tube, Conical Bottom, with Dome Seal Screw Cap, Sterile, 50/Rack, 500/Case 4846029 Mfr # 352099
COMPARE
50/Rack, 500/Case 15
₹76,000.00INR / Case of 500
Falcon 15 mL Polystyrene Centrifuge Tube, Conical Bottom, with Dome Seal Screw Cap, Sterile, 50/Bag 4846059 Mfr # 352095
COMPARE
50/Bag 15
₹8,400.00INR / Pkg of 50
Falcon 15 mL Polystyrene Centrifuge Tube, Conical Bottom, with Dome Seal Screw Cap, Sterile, 50/Rack 4846060 Mfr # 352099
COMPARE
50/Rack 15
₹8,800.00INR / Pkg of 50