Falcon® High-Clarity Polypropylene (PP) Centrifuge Tubes, Conical Bottom
Built for strength, clarity, and sample protection
  • Durable polypropylene construction supports high-stress centrifugation, vortexing, and long-term storage
  • Unique double-threaded cap design helps reduce cross-threading and leakage for more secure handling
  • High-clarity tubes with dark blue graduations and white marking areas improve readability and sample identification
  • Designed to meet bioanalytical-grade standards with low protein binding and consistent performance
Falcon® conical centrifuge tubes are engineered to deliver reliable performance across a wide range of laboratory applications. Constructed from high-strength polypropylene, these tubes are designed to withstand demanding conditions, including high-speed centrifugation, vortexing, and long-term storage at temperatures as low as -80°C. A unique double-threaded cap design enhances sealing integrity while reducing the risk of cross-threading, helping protect valuable samples during handling and transport. High-clarity construction, combined with dark blue graduations and a dedicated white marking area, improves visibility and labeling accuracy. Available in multiple sizes and packaging formats, Falcon tubes provide a dependable solution for both routine and critical research workflows.
8,600.00 - ₹103,000.00INR / Pkg of 50
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Style Packaging Tube Capacity (mL)
Availability Pricing
Falcon 15 mL High Clarity PP Centrifuge Tube, Conical Bottom, with Dome Seal Screw Cap, Sterile, 50/Bag, 500/Case 4846025 Mfr # 352096
COMPARE
Dome seal screw cap 50/Bag, 500/Case 15
₹74,100.00INR / Case of 500
Falcon 15 mL High Clarity PP Centrifuge Tube, Conical Bottom, with Dome Seal Screw Cap and Rack, Sterile, 50/Bag, 500/Case 4846026 Mfr # 352196
COMPARE
Dome seal screw cap 50/Bag, 500/Case, Rack Included 15
₹73,700.00INR / Case of 500
Falcon 15 mL High Clarity PP Centrifuge Tube, Conical Bottom, with Dome Seal Screw Cap, Sterile, 50/Rack, 500/Case 4846027 Mfr # 352097
COMPARE
Dome seal screw cap 50/Rack, 500/Case 15
₹76,400.00INR / Case of 500
Falcon 50 mL High Clarity PP Centrifuge Tube, Conical Bottom, with Flat Top Cap, Sterile, 25/Bag, 500/Case 4846030 Mfr # 352070
COMPARE
Flat top 25/Bag, 500/Case 50
₹99,800.00INR / Case of 500
Falcon 50 mL High Clarity PP Centrifuge Tube, Conical Bottom, with Flat Top Cap, Sterile, 25/Rack, 500/Case 4846031 Mfr # 352098
COMPARE
Flat top 25/Rack, 500/Case 50
₹103,000.00INR / Case of 500
Falcon 15 mL High Clarity PP Centrifuge Tube, Conical Bottom, with Dome Seal Screw Cap, Sterile, 50/Bag 4846056 Mfr # 352096
COMPARE
Dome seal screw cap 50/Bag 15
₹8,600.00INR / Pkg of 50
Falcon 15 mL High Clarity PP Centrifuge Tube, Conical Bottom, with Dome Seal Screw Cap and Rack, Sterile, 50/Bag 4846057 Mfr # 352196
COMPARE
Dome seal screw cap 50/Bag, Rack Included 15
₹8,600.00INR / Pkg of 50
Falcon 15 mL High Clarity PP Centrifuge Tube, Conical Bottom, with Dome Seal Screw Cap, Sterile, 50/Rack 4846058 Mfr # 352097
COMPARE
Dome seal screw cap 50/Rack 15
₹8,800.00INR / Pkg of 50
Falcon 50 mL High Clarity PP Centrifuge Tube, Conical Bottom, with Flat Top Cap, Sterile, 25/Bag 4846061 Mfr # 352070
COMPARE
Flat top 25/Bag 50
₹11,200.00INR / Pkg of 25
Falcon 50 mL High Clarity PP Centrifuge Tube, Conical Bottom, with Flat Top Cap, Sterile, 25/Rack 4846062 Mfr # 352098
COMPARE
Flat top 25/Rack 50
₹11,600.00INR / Pkg of 25
Showing 1- 10 of 10