Extech Sound Meter with RS-232
Background-noise absorber filters ambient sounds for accurate machine-noise measurements
Meter features six sound level ranges, analog bar graph, and RS-232 interface for capturing data directly on your PC (order software separately). Use the selectable backlit display for easy readability in low kight conditions. What''s included: microphone windscreen, carrying case, and one 9 V battery. Compliance: ANSI S1.4-1983 Type 2, IEC 651 Type 2
92,200.00 - ₹145,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Includes
Availability Pricing
Extech 407750 Integrating Sound Meter with RS-232 4042500 Mfr # 407750
COMPARE
Meter only
₹92,200.00INR / Each
Extech 407750-NIST Integrating Sound Meter with RS-232 and NIST-Traceable Calibration 4042510 Mfr # 407750-NIST
COMPARE
Meter with NIST-Traceable calibration
₹145,000.00INR / Each