Extech Receptacle Testers
Safely test electrical sockets
  • Lights indicate circuit condition
  • Test for correct wiring of three-wire receptacles in 110 to 125 VAC, 50/60 Hz circuits
  • Detect five wiring faults: Open Ground, Open Neutral, Open Hot, Hot/Ground Reverse, Hot/Neutral Reverse
Quickly and safely test for faulty wiring in Type B electrical outlets with the receptacle testers. To use, simply plug the tester into a standard North American three-wire receptacle. Testers have indicator lights that light up instantly when faulty wiring is detected; lights also verify correctly wired receptacle. A test code light legend on the tester provides a quick and easy identification of potential hazards.
1,700.00 - ₹2,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Measurement Safety Rating Description
Availability Pricing
Extech ET11 GFCI Receptacle Tester with Pull Bar 3975923 Mfr # ET11
COMPARE
Open ground, open neutral, open hot, hot & ground reversed, hot & neutral reversed,correct wiring C-UL-US GFCI Receptacle Tester with Pull Bar
₹2,800.00INR / Each
Extech ET16 Receptacle Tester with Pull Bar 3975924 Mfr # ET16
COMPARE
Receptacle Tester with Pull Bar
₹1,700.00INR / Each