Extech Precision Thermohygrometer with Dew Point and Wet Bulb
Measures relative humidity with ±2% accuracy
Precision capacitance humidity sensor offers fast response. Slim design with rubberized sides for better grip and easy, one-handed operation. The dual LCD provides simultaneous display of humidity and temperature, or humidity and dew point, or humidity and wet bulb. Advanced features include data hold, min/max readings, low-battery indication, and auto power off.
45,500.00 - ₹95,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RH Range (%) Max Temperature (° F) Description
Availability Pricing
Extech RH390 Precision Thermohygrometer, 0 to 100% RH/, -22 to 199F WW-37803-33 Mfr # RH390
COMPARE
100 199 Precision Thermohygrometer, 0 to 100% RH/, -22 to 199F
₹45,500.00INR / Each
Extech RH490 Precision Psychrometer (GPP, g/kg) WW-37851-01 Mfr # RH490
COMPARE
100 199 Precision Psychrometer (GPP, g/kg)
₹95,600.00INR / Each