Extech Multifunction Environmental Thermoanemometers
Measures 11 environmental conditions including light
  • Auto-rotating, dual-line LCD with backlight for easy reading in low light
  • Operational modes include auto-off and low-battery indicators, MIN/MAX, and data hold
Versatile anemometer uses a low-friction vane and displays air velocity measurements in five units. High-precision humidity sensor provides a quick response time to make accurate calculations for wet bulb, dew point, heat index and windchill readings. Dual-line display simultaneously shows relative humidity and temperature, or air velocity and temperature. Optional external high-accuracy temperature probe expands the measurement range to 14 to 212°F (-10 to 100°C).
65,000.00 - ₹108,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
Extech 45170 4 in 1 Environmental Thermoanemometer 1030002 Mfr # 45170
COMPARE
±3% full scale 10 ft/min 5910
₹65,000.00INR / Each
Extech EN300 5 in 1 Environmental Thermoanemometer 1030003 Mfr # EN300
COMPARE
±3% full scale 1 ft/min 5910
₹108,000.00INR / Each