Extech Low-Pressure Differential Manometer
Low range and high resolution in gauge or differential pressure measurement
  • Measure from 0 to 55.4'' WC
  • Very large LCD with backlight
Designed for use in cleanrooms, mold remediation, animal physiology, and general HVAC applications, this durable, easy-to-use manometer features 11 units of measure, min/max recording, relative time stamp, and built-in USB port. Functions include data hold, auto power off, min/max functions, and a zero function for offset correction or measurement.
55,100.00 - ₹57,500.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (PSID) Resolution
Availability Pricing
Extech HD755 Heavy-Duty Differential Pressure Manometer, 0.5 psi WW-68602-01 Mfr # HD755
COMPARE
±0.3% full-scale 0.5 0.001 psi
₹55,100.00INR / Each
Extech HD700 Heavy-Duty Differential Pressure Manometer, +/-2 psi WW-68602-02 Mfr # HD700
COMPARE
±0.3% full-scale 2 0.001 psi
₹55,100.00INR / Each
Extech HD750 Heavy-Duty Differential Pressure Manometer, 5 psi WW-68602-03 Mfr # HD750
COMPARE
±0.3% full-scale 5 0.001 psi
₹57,500.00INR / Each