Extech Low-Cost Digital Force Gauges
Economical testing solution for lab or field
These force gauges are ideal for a variety of applications in the electronics, chemical, and plastics industries. Gauges feature a five-digit LCD with reversible display to match viewing angle; selectable fast/slow response mode; overrange, low battery indication; zero adjust push button; and peak hold switch.
134,000.00 - ₹173,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity
Availability Pricing
Extech 475040 Gauge, 176 oz., 5000 g, 49 N 2530408 Mfr # 475040
COMPARE
176 oz
₹134,000.00INR / Each
Extech 475044 Gauge, 44 lb., 20 kg, 196 N 2530409 Mfr # 475044
COMPARE
44 lbs
₹173,000.00INR / Each