Extech Heavy-Duty Hot-Wire Thermoanemometer with PC Interface
Accurately measures airflow (cfm) or velocity plus temperature
  • Measure airflow down to 40 ft/min
  • Ideal for measuring in HVAC ducts and other small vents
  • Super-large 1.4" (36 mm) dual LCD display
  • Min/Max, Data Hold
  • Optional RS-232 interface and data acquisition software (10200-10) enables user to display and capture readings on a PC, and set time intervals
  • Optional datalogger (10200-20) allows user to capture up to 8000 readings and can be transferred to a PC
This hot-wire anemometer displays air velocity and temperature or CFM/CMM and temperature simultaneously on the large, dual LCD. Average up to 20 air volume points for a mean calculation. Other features include built-in RS-232 serial port, min/max, data hold, and auto shut-off. Meter is ideal for duct and ventilation measurements. Increase logging capabilities to 8000 data points with data module 10200-20 (sold separately). Download data to a PC via RS-232 interface (sold separately).
147,000.00 - ₹174,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Air Velocity Accuracy Air Velocity Resolution Max Air Velocity (ft/min)
Availability Pricing
Extech 407123 Heavy-Duty Hot-Wire Thermoanemometer with PC Interface 1020002 Mfr # 407123
COMPARE
±3% 10 ft/min (0.1 m/s, 0.1 mph, 0.1 km/hr, 0.1 knots) 3940
₹147,000.00INR / Each
Extech 407119 Heavy-Duty CFM Hot-Wire Thermoanemometer with PC Interface 1020009 Mfr # 407119
COMPARE
±(5% of reading + 5 digits) 1 ft/min (0.1 m/s, 0.1 mph, 0.1 knots) 3346
₹174,000.00INR / Each