Extech EX500 Series True-RMS Heavy-Duty Industrial Multimeters
Durable true-rms multimeters have 1000 V input protection on all functions
  • True rms for reliable measurements on non-sine wave forms
  • 1000 V input protection on all functions
  • IP67-rated, waterproof and dustproof housing
  • 2 m (6.5 ft) drop-tested
  • Accepts a type K thermocouple probe to measure up to 760 °C (1400 °F)*
These heavy-duty multimeters feature IP67-rated housing and withstand a 2 m (6.5 ft) drop, making them ideal for demanding work environments. Meters measure AC/DC voltage up to 1000 V, AC/DC current, resistance, capacitance, frequency, duty cycle, temperature*, diode, and continuity.
30,000.00 - ₹56,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) Ac Voltage Accuracy Resistance Max (MΩ)
Availability Pricing
Extech EX505A True-RMS Heavy-Duty Industrial Multimeter 3975925 Mfr # EX505A
COMPARE
1000 60 mV range: ±(0.9% of reading + 9 digits)<br>600 mV, 6 V, 60 V, and 600 V ranges: ±(0.8% of reading + 3 digits)<br>1000 V range: ±(0.8% of reading + 8 digits) 60
₹30,000.00INR / Each
Extech EX505A-K True-RMS Heavy-Duty Industrial Multimeter Kit 3975926 Mfr # EX505A-K
COMPARE
1000 60 mV range: ±(0.9% of reading + 9 digits)<br>600 mV, 6 V, 60 V, and 600 V ranges: ±(0.8% of reading + 3 digits)<br>1000 V range: ±(0.8% of reading + 8 digits) 60
₹39,400.00INR / Each
Extech EX520A High-Accuracy True-RMS Heavy-Duty Industrial Multimeter with LoZ and Low-Pass Filter 3975927 Mfr # EX520A
COMPARE
1000 6, 60, and 600 V ranges: ±(1.0% of reading + 3 digits)<br>1000 V range: ±(1.0% of reading + 8 digits) 40
₹42,900.00INR / Each
Extech EX530A High-Precision True-RMS Heavy-Duty Industrial Multimeter with LoZ and Low-Pass Filter 3975928 Mfr # EX530A
COMPARE
1000 500 mV, 5 V, 50 V, and 500 V ranges: ±(1.0% of reading + 4 digits)<br>1000 V range: ±(1.0% of reading + 3 digits) 50
₹56,900.00INR / Each