Extech EasyView CFM/CMM Vane Thermoanemometers
Compact and rugged design featuring a large display
  • View measurements in low light areas with backlit display
  • Handheld air vane with 4-foot (1.2 m) cable length offers greater measurement flexibility
  • Simultaneous display of ambient temperature and airflow or air velocity
  • Up to 8 easy-to-set area dimensions (ft2 or cm2) are stored in the meter’s internal memory
  • 20 point average function for airflow
These thermoanemometers are perfect for use in lab or plant maintenance applications, environmental analysis, fume hood testing, and HVAC applications. Each meter simultaneously displays air volume (CFM), air velocity and ambient temperature (°F or °C) on the large backlit display.
Additional features include: 20 point air flow averaging, Data Hold, Min/Max, and auto-power-off. User can select desired unit of measure for both temperature and air velocity.
60,900.00 - ₹78,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Air Volume (CFM) Volume Resolution Description
Availability Pricing
Extech AN100 EasyView CFM/CMM Vane Thermoanemometer, 1036032 Mfr # AN100
COMPARE
999900 0.1 CFM EasyView CFM/CMM Vane Thermoanemometer,
₹60,900.00INR / Each
Extech AN200 EasyView CFM/CMM Vane Thermoanemometer with IR Thermometer, 1037996 Mfr # AN200
COMPARE
999999 0.1 CFM EasyView CFM/CMM Vane Thermoanemometer with IR Thermometer,
₹78,600.00INR / Each