Extech® DT Series Laser Distance Meters
Automatically calculates area and volume
  • Store up to 20 data points
  • Automatic shutoff improves battery life
  • Continuous measurement with Min/Max distance tracking
The DT series laser distance meter quickly and accurately measures distance and calculates area, volume, and indirect readings using the integrated Pythagorean theorem with just the push of a button. The sum and differences of multiple readings are easily visible on the large quad backlit LCD. Features include front or rear edge reference, laser pointer, stakeout function, built-in bubble level, low-battery indicator, and double-molded housing.
17,700.00 - ₹36,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (mm) Max range (m) Resolution
Availability Pricing
Extech DT40M Laser Distance Meter, 40 M 3550000 Mfr # DT40M
COMPARE
±2 40 0.0", 0.00 ft, 0.000 m
₹17,700.00INR / Each
Extech DT60M Laser Distance Meter, 60 M 3550001 Mfr # DT60M
COMPARE
±2 60 0.0", 0.00 ft, 0.000 m
₹23,600.00INR / Each
Extech DT100M Laser Distance Meter, 100 M 3550002 Mfr # DT100M
COMPARE
±2 100 0.0", 0.00 ft, 0.000 m
₹36,300.00INR / Each